Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-chloro-4-hydroxybenzoate, 98%

$78.04 - $982.05

Chemical Identifiers

Chemical Name or Material Methyl 3-chloro-4-hydroxybenzoate
CAS 3964-57-6
Molecular Formula C8H7ClO3
Formula Weight 186.6
MDL Number MFCD00016418
Beilstein 2641291
Synonym 3-Chloro-4-hydroxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1238914

  SDS
alfa aesar™
A1238914
25g Each for $292.46

AAA1238906

  SDS
alfa aesar™
A1238906
5g Each for $78.04

AAA1238922

  SDS
alfa aesar™
A1238922
100g Each for $982.05
Description & Specifications

Specifications

Formula Weight 186.6
Chemical Name or Material Methyl 3-chloro-4-hydroxybenzoate
Assay 98%
Molecular Formula C8H7ClO3
MDL Number MFCD00016418
CAS 3964-57-6
Beilstein 2641291
Synonym 3-Chloro-4-hydroxybenzoic acid methyl ester
Health Hazard 1 Warning
Melting Point 104° to 109°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.