missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-bromobenzenesulfonate, 97%

Chemical Identifiers

CAS 1421261-59-7
Molecular Formula C7H7BrO3S
Molecular Weight (g/mol) 251.094
MDL Number MFCD22683148
InChI Key RBLPSOBJNBVDMD-UHFFFAOYSA-N
Synonym 3-Bromobenzenesulfonic acid methyl ester
PubChem CID 21222348
IUPAC Name methyl 3-bromobenzenesulfonate
SMILES COS(=O)(=O)C1=CC(=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60814MD View Documents Alfa Aesar™
H60814MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6081403 View Documents Alfa Aesar™
H6081403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421261-59-7
251.094
RBLPSOBJNBVDMD-UHFFFAOYSA-N
21222348
COS(=O)(=O)C1=CC(=CC=C1)Br
C7H7BrO3S
MFCD22683148
3-Bromobenzenesulfonic acid methyl ester
methyl 3-bromobenzenesulfonate

Specifications

250mg
C7H7BrO3S
3-Bromobenzenesulfonic acid methyl ester
COS(=O)(=O)C1=CC(=CC=C1)Br
251.094
251.09
Methyl 3-bromobenzenesulfonate
1421261-59-7
MFCD22683148
RBLPSOBJNBVDMD-UHFFFAOYSA-N
methyl 3-bromobenzenesulfonate
21222348
97%