Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-bromo-4-(hydroxymethyl)benzoate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 3-bromo-4-(hydroxymethyl)benzoate
CAS 90326-62-8
Formula Weight 245.07
MDL Number MFCD15527253
Beilstein 2642910
Synonym 3-Bromo-4-(hydroxymethyl)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6687603

 
alfa aesar™
H6687603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66876MD

 
alfa aesar™
H66876MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.07
Chemical Name or Material Methyl 3-bromo-4-(hydroxymethyl)benzoate
Physical Form Solid
MDL Number MFCD15527253
CAS 90326-62-8
Assay 97%
Synonym 3-Bromo-4-(hydroxymethyl)benzoic acid methyl ester
Quantity 1g
Beilstein 2642910