Learn More
Methyl 3-bromo-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3022218
Description
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Methyl 3-bromo-4-hydroxybenzoate | |
108°C to 110°C | |
MFCD06203850 | |
methyl3-bromo-4-hydroxybenzoate, 3-bromo-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-bromo-4-hydroxy-, methyl ester, 2-bromo-4-methoxycarbonyl phenol, methyl 4-hydroxy-3-bromobenzoate, methyl-3-bromo-4-hydroxybenzoate, methyl 3-bromo-4-hydroxy-benzoate, methyl 3-bromo-4-hydroxybenzoate, methyl 3-bromanyl-4-oxidanyl-benzoate, 3-bromo-4-hydroxy-benzoic acid methyl ester | |
COC(=O)C1=CC(Br)=C(O)C=C1 | |
231.05 | |
231.05 |
29415-97-2 | |
C8H7BrO3 | |
50 g | |
RKUNSPWAQIUGEZ-UHFFFAOYSA-N | |
methyl 3-bromo-4-hydroxybenzoate | |
4778958 | |
98% |
Chemical Identifiers
29415-97-2 | |
231.05 | |
RKUNSPWAQIUGEZ-UHFFFAOYSA-N | |
4778958 | |
COC(=O)C1=CC(Br)=C(O)C=C1 |
C8H7BrO3 | |
MFCD06203850 | |
methyl3-bromo-4-hydroxybenzoate, 3-bromo-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-bromo-4-hydroxy-, methyl ester, 2-bromo-4-methoxycarbonyl phenol, methyl 4-hydroxy-3-bromobenzoate, methyl-3-bromo-4-hydroxybenzoate, methyl 3-bromo-4-hydroxy-benzoate, methyl 3-bromo-4-hydroxybenzoate, methyl 3-bromanyl-4-oxidanyl-benzoate, 3-bromo-4-hydroxy-benzoic acid methyl ester | |
methyl 3-bromo-4-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only