missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Bromo-2-methylbenzoate, TCI America™

$171.11 - $513.34

Chemical Identifiers

CAS 99548-54-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD09031772
InChI Key MKQQTCSBXHAYQL-UHFFFAOYSA-N
Synonym 3-Bromo-2-methylbenzoic Acid Methyl Ester, Methyl 3-Bromo-o-toluate, 3-Bromo-o-toluic Acid Methyl Ester
PubChem CID 12929135
IUPAC Name methyl 3-bromo-2-methylbenzoate
SMILES CC1=C(C=CC=C1Br)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25435G View Documents TCI America
M25435G
5g Each for $171.11
 
M254325G View Documents TCI America
M254325G
25g Each for $513.34
 
Specifications

Chemical Identifiers

99548-54-6
229.073
MKQQTCSBXHAYQL-UHFFFAOYSA-N
12929135
CC1=C(C=CC=C1Br)C(=O)OC
C9H9BrO2
MFCD09031772
3-Bromo-2-methylbenzoic Acid Methyl Ester, Methyl 3-Bromo-o-toluate, 3-Bromo-o-toluic Acid Methyl Ester
methyl 3-bromo-2-methylbenzoate

Specifications

140°C
32°C
99548-54-6
MFCD09031772
MKQQTCSBXHAYQL-UHFFFAOYSA-N
methyl 3-bromo-2-methylbenzoate
12929135
Crystal-Lump at 20°C
Methyl 3-Bromo-2-methylbenzoate
White-Yellow
5g
C9H9BrO2
3-Bromo-2-methylbenzoic Acid Methyl Ester, Methyl 3-Bromo-o-toluate, 3-Bromo-o-toluic Acid Methyl Ester
CC1=C(C=CC=C1Br)C(=O)OC
229.073
229.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No