missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-bromo-2-fluorobenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3686503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 3-bromo-2-fluorobenzoate | |
35°C to 37°C | |
C8H6BrFO2 | |
1 g | |
ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
methyl 3-bromo-2-fluorobenzoate | |
21584505 | |
98% |
206551-41-9 | |
White to Cream | |
MFCD09261255 | |
3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
COC(=O)C1=C(C(=CC=C1)Br)F | |
233.036 | |
233.04 | |
Fused Solid |
Chemical Identifiers
206551-41-9 | |
233.036 | |
ZWOFHFOFKBYRHV-UHFFFAOYSA-N | |
21584505 | |
COC(=O)C1=C(C(=CC=C1)Br)F |
C8H6BrFO2 | |
MFCD09261255 | |
3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester | |
methyl 3-bromo-2-fluorobenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only