Please login to your online account to display your discounted pricing

Methyl 3-bromo-2-(bromomethyl)propionate, 97%, ACROS Organics™

$114.79 - $114.79

Chemical Identifiers

Chemical Name or Material Methyl 3-bromo-2-(bromomethyl)propionate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H8Br2O2
Linear Formula BrCH2CH(CH2Br)CO2C2H5
Formula Weight 259.92
MDL Number MFCD00011534
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373420010

  SDS
thermo fisher scientific
AC373420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373420050

  SDS
thermo fisher scientific
AC373420050
Glass bottle 5g Each for $114.79
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 259.92
Quantity 1g
Color Clear colorless to yellow
Density 1.8200g/mL
Refractive Index 1.5070 to 1.5100 (20°C, 589nm)
Linear Formula BrCH2CH(CH2Br)CO2C2H5
Chemical Name or Material Methyl 3-bromo-2-(bromomethyl)propionate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 22262-60-8
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 97%
Molecular Formula C5H8Br2O2
MDL Number MFCD00011534
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.