Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-benzoylpropionate, 98%

$85.43 - $361.62

Chemical Identifiers

Chemical Name or Material Methyl 3-benzoylpropionate
CAS 25333-24-8
Molecular Formula C11H12O3
Formula Weight 192.22
MDL Number MFCD00014906
Beilstein 2211627
Synonym 3-Benzoylpropionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2043506

  SDS
alfa aesar™
B2043506
5g Each for $85.43

AAB2043514

  SDS
alfa aesar™
B2043514
25g Each for $361.62
Description & Specifications

Specifications

Formula Weight 192.22
Chemical Name or Material Methyl 3-benzoylpropionate
Assay 98%
Molecular Formula C11H12O3
MDL Number MFCD00014906
CAS 25333-24-8
Beilstein 2211627
Synonym 3-Benzoylpropionic acid methyl ester
Boiling Point 172° to 174°C (10mmHg)
Quantity 5g

Applications
Methyl 3-benzoylpropionate is used as a chemical and organic intermediate.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.