Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-aminopyrazine-2-carboxylate, 99%

$59.82 - $289.30

Chemical Identifiers

Chemical Name or Material Methyl 3-aminopyrazine-2-carboxylate
CAS 16298-03-6
Molecular Formula C6H7N3O2
Formula Weight 153.14
MDL Number MFCD00010097
Synonym 3-Aminopyrazine-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH2917203

  SDS
alfa aesar™
H2917203
Each for $59.82

AAH2917209

  SDS
alfa aesar™
H2917209
Each for $289.30
Description & Specifications

Specifications

Formula Weight 153.14
Chemical Name or Material Methyl 3-aminopyrazine-2-carboxylate
CAS 16298-03-6
Melting Point 171° to 174°C
MDL Number MFCD00010097
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Aminopyrazine-2-carboxylic acid methyl ester
Health Hazard 1 Warning
Molecular Formula C6H7N3O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.