missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-aminocrotonate, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 156795000

 View more versions of this product

Catalog No. AC156795000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 3-aminocrotonate
Authentic
1% max. (K.F.)
81.0°C to 83.0°C
97%
97%
CH3C(NH2)=CHCO2CH3
03, 632
Solubility in water: insoluble in water. Other solubilities: solubility in chloroform >500g/L (20°C),solubility in acetone >500g/L (20°C)
CC(=CC(=O)OC)N
115.13
115.13
97%
91°C
Plastic bottle
White to Yellow
500g
14205-39-1
C5H9NO2
MFCD00071534
3-amino-2-butenoic acid methyl ester
XKORCTIIRYKLLG-ARJAWSKDSA-N
methyl (Z)-3-aminobut-2-enoate
643918
Crystalline Powder

Chemical Identifiers

14205-39-1
115.13
XKORCTIIRYKLLG-ARJAWSKDSA-N
643918
CC(=CC(=O)OC)N
C5H9NO2
MFCD00071534
3-amino-2-butenoic acid methyl ester
methyl (Z)-3-aminobut-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-056-5

RTECSNumber : EM9092500

TSCA : TSCA

SDS Documents