Learn More
Methyl 3-aminocrotonate, 97%, Thermo Scientific Chemicals
$56.42 - $162.38
Chemical Identifiers
CAS | 14205-39-1 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.13 |
MDL Number | MFCD00008072,MFCD00008072,MFCD00008072 |
InChI Key | XKORCTIIRYKLLG-ONEGZZNKSA-N |
Synonym | methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester |
PubChem CID | 643918 |
IUPAC Name | methyl (Z)-3-aminobut-2-enoate |
SMILES | COC(=O)\C=C(/C)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2495322
|
Thermo Scientific Chemicals
B2495322 |
100 g |
Each for $56.42
|
|
|||||
AAB2495336
|
Thermo Scientific Chemicals
B2495336 |
500 g |
Each for $162.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14205-39-1 | |
115.13 | |
XKORCTIIRYKLLG-ONEGZZNKSA-N | |
643918 | |
COC(=O)\C=C(/C)N |
C5H9NO2 | |
MFCD00008072,MFCD00008072,MFCD00008072 | |
methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester | |
methyl (Z)-3-aminobut-2-enoate |
Specifications
14205-39-1 | |
122°C (42 mmHg) | |
C5H9NO2 | |
100 g | |
Hygroscopic | |
XKORCTIIRYKLLG-ONEGZZNKSA-N | |
methyl (Z)-3-aminobut-2-enoate | |
643918 | |
97% |
81°C to 85°C | |
91°C (195°F) | |
MFCD00008072,MFCD00008072,MFCD00008072 | |
956592 | |
methyl 3-aminocrotonate, methyl 3-aminobut-2-enoate, methyl 2z-3-aminobut-2-enoate, z-methyl 3-aminobut-2-enoate, 3-aminocrotonic acid methyl ester, methyl beta-aminocrotonate, methyl z-3-aminobut-2-enoate, methyl 2z-3-amino-2-butenoate, beta-aminocrotonic acid methyl ester, 3-amino-2-butenoic acid methyl ester | |
COC(=O)\C=C(/C)N | |
115.13 | |
115.13 | |
Methyl 3-aminocrotonate |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H317-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 238-056-5
RTECSNumber : EM9092500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only