Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-6-iodopyrazine-2-carboxylate, 96%

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-6-iodopyrazine-2-carboxylate
CAS 1458-16-8
Formula Weight 279.04
MDL Number MFCD09909640
Synonym 3-Amino-6-iodopyrazine-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6302603

 
alfa aesar™
H6302603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63026MD

 
alfa aesar™
H63026MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6302606

 
alfa aesar™
H6302606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 279.04
Chemical Name or Material Methyl 3-amino-6-iodopyrazine-2-carboxylate
Sensitivity Light Sensitive
Physical Form Solid
Assay 96%
MDL Number MFCD09909640
CAS 1458-16-8
Synonym 3-Amino-6-iodopyrazine-2-carboxylic acid methyl ester
Quantity 1g