missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-6-iodopyrazine-2-carboxylate, 96%

Manufacturer:  Alfa Aesar™ H6302606

 View more versions of this product

Catalog No. AAH6302606



Specifications

Specifications

Methyl 3-amino-6-iodopyrazine-2-carboxylate
5g
C6H6IN3O2
3-amino-6-iodopyrazine-2-carboxylic acid methyl ester, 3-amino-6-iodo-pyrazine-2-carboxylic acid methyl ester, methyl 3-amino-6-iodo-2-pyrazinecarboxylate, methyl 3-azanyl-6-iodanyl-pyrazine-2-carboxylate, 3-amino-6-iodo-2-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid,3-amino-6-iodo-,methyl ester, 2-pyrazinecarboxylicacid, 3-amino-6-iodo-, methyl ester
COC(=O)C1=NC(=CN=C1N)I
279.037
279.04
96%
Light Sensitive
1458-16-8
MFCD09909640
FDLARAKNPMCCKJ-UHFFFAOYSA-N
methyl 3-amino-6-iodopyrazine-2-carboxylate
11087124
Solid

Chemical Identifiers

1458-16-8
279.037
FDLARAKNPMCCKJ-UHFFFAOYSA-N
11087124
COC(=O)C1=NC(=CN=C1N)I
C6H6IN3O2
MFCD09909640
3-amino-6-iodopyrazine-2-carboxylic acid methyl ester, 3-amino-6-iodo-pyrazine-2-carboxylic acid methyl ester, methyl 3-amino-6-iodo-2-pyrazinecarboxylate, methyl 3-azanyl-6-iodanyl-pyrazine-2-carboxylate, 3-amino-6-iodo-2-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid,3-amino-6-iodo-,methyl ester, 2-pyrazinecarboxylicacid, 3-amino-6-iodo-, methyl ester
methyl 3-amino-6-iodopyrazine-2-carboxylate
Documents