Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-6-chloropyrazine-2-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-6-chloropyrazine-2-carboxylate
CAS 1458-03-3
Formula Weight 187.58
MDL Number MFCD00834965
Synonym 3-Amino-6-chloropyrazine-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6325003

 
alfa aesar™
H6325003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6325006

 
alfa aesar™
H6325006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63250MD

 
alfa aesar™
H63250MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.58
Synonym 3-Amino-6-chloropyrazine-2-carboxylic acid methyl ester
Chemical Name or Material Methyl 3-amino-6-chloropyrazine-2-carboxylate
MDL Number MFCD00834965
Physical Form Solid
Assay 97%
Quantity 1g
CAS 1458-03-3