missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-6-chloropyrazine-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6325006

 View more versions of this product

Catalog No. AAH6325006



Specifications

Specifications

Methyl 3-amino-6-chloropyrazine-2-carboxylate
1458-03-3
MFCD00834965
JGAJCAJHSHUPGH-UHFFFAOYSA-N
methyl 3-amino-6-chloropyrazine-2-carboxylate
824637
Solid
5g
C6H6ClN3O2
methyl 2-amino-5-chloropyrazine-3-carboxylate, methyl 3-amino-6-chloro-2-pyrazinecarboxylate, 3-amino-6-chloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-6-chloro-, methyl ester, methyl3-amino-6-chloropyrazine-2-carboxylate, ksc524o0p, methyl 2-amino-5-chloro-3-pyrazinylcarboxylate, methyl 3-amino-6-chloro-pyrazine-2-carboxylate, methyl 3-amino-6-chloropyrazinecarboxylate
COC(=O)C1=NC(=CN=C1N)Cl
187.583
187.58
97%

Chemical Identifiers

1458-03-3
187.583
JGAJCAJHSHUPGH-UHFFFAOYSA-N
824637
COC(=O)C1=NC(=CN=C1N)Cl
C6H6ClN3O2
MFCD00834965
methyl 2-amino-5-chloropyrazine-3-carboxylate, methyl 3-amino-6-chloro-2-pyrazinecarboxylate, 3-amino-6-chloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-6-chloro-, methyl ester, methyl3-amino-6-chloropyrazine-2-carboxylate, ksc524o0p, methyl 2-amino-5-chloro-3-pyrazinylcarboxylate, methyl 3-amino-6-chloro-pyrazine-2-carboxylate, methyl 3-amino-6-chloropyrazinecarboxylate
methyl 3-amino-6-chloropyrazine-2-carboxylate
Documents