missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Amino-6-bromopyrazine-2-carboxylate, TCI America™

$76.39 - $297.92

Chemical Identifiers

CAS 6966-01-4
Molecular Formula C6H6BrN3O2
Molecular Weight (g/mol) 232.04
MDL Number MFCD00834964
InChI Key CNXSIRHOIFRMOB-UHFFFAOYSA-N
Synonym methyl3-amino-6-bromopyrazine-2-carboxylate, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 2-amino-5-bromo-3-methoxycarbonyl pyrazine, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, pubchem22386
PubChem CID 227251
IUPAC Name methyl 3-amino-6-bromopyrazine-2-carboxylate
SMILES COC(=O)C1=NC(=CN=C1N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26555G View Documents TCI America
M26555G
5g Each for $297.92
 
M26551G View Documents TCI America
M26551G
1g Each for $76.39
 
Specifications

Chemical Identifiers

6966-01-4
232.04
CNXSIRHOIFRMOB-UHFFFAOYSA-N
227251
COC(=O)C1=NC(=CN=C1N)Br
C6H6BrN3O2
MFCD00834964
methyl3-amino-6-bromopyrazine-2-carboxylate, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 2-amino-5-bromo-3-methoxycarbonyl pyrazine, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, pubchem22386
methyl 3-amino-6-bromopyrazine-2-carboxylate

Specifications

Undesignated
5g
C6H6BrN3O2
methyl3-amino-6-bromopyrazine-2-carboxylate, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 2-amino-5-bromo-3-methoxycarbonyl pyrazine, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, pubchem22386
COC(=O)C1=NC(=CN=C1N)Br
232.04
232.04
≥98.0% (GC)
174°C
6966-01-4
MFCD00834964
CNXSIRHOIFRMOB-UHFFFAOYSA-N
methyl 3-amino-6-bromopyrazine-2-carboxylate
227251
Crystal-Powder at 20°C
Methyl 3-Amino-6-bromopyrazine-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No