missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-6-bromopyrazine-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H63537MD

 View more versions of this product

Catalog No. AAH63537MD



Specifications

Specifications

Methyl 3-amino-6-bromopyrazine-2-carboxylate
172°C to 177°C
6966-01-4
MFCD00834964
CNXSIRHOIFRMOB-UHFFFAOYSA-N
methyl 3-amino-6-bromopyrazine-2-carboxylate
227251
Solid
White to Yellow
250mg
C6H6BrN3O2
2-amino-5-bromo-3-methoxycarbonyl pyrazine, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl3-amino-6-bromopyrazine-2-carboxylate, pubchem22386, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester
COC(=O)C1=NC(=CN=C1N)Br
232.04
232.03
97%

Chemical Identifiers

6966-01-4
232.04
CNXSIRHOIFRMOB-UHFFFAOYSA-N
227251
COC(=O)C1=NC(=CN=C1N)Br
C6H6BrN3O2
MFCD00834964
2-amino-5-bromo-3-methoxycarbonyl pyrazine, 2-amino-5-bromopyrazine-3-carboxylic acid methyl ester, 3-amino-6-bromopyrazine-2-carboxylic acid methyl ester, 5-amino-2-bromo-6-pyrazinecarboxylic acidmethyl ester, methyl 3-amino-6-bromo-2-pyrazinecarboxylate, methyl 3-amino-6-bromo-pyrazine-2-carboxylate, methyl 6-bromo-3-amino-2-pyrazinecarboxylate, methyl3-amino-6-bromopyrazine-2-carboxylate, pubchem22386, pyrazinecarboxylic acid, 3-amino-6-bromo-, methyl ester
methyl 3-amino-6-bromopyrazine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents