Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate, 97%

$398.27 - $398.27

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
CAS 175137-08-3
Molecular Formula C12H10FNO2S
Formula Weight 251.28g/mol
MDL Number MFCD00068133
Synonym 3-Amino-5-(4-fluorophenyl)thiophene-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5035406

 
alfa aesar™
H50354-06
5g Each for $398.27
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate
CAS 175137-08-3
Assay 97%
Molecular Formula C12H10FNO2S
Formula Weight 251.28g/mol
MDL Number MFCD00068133
Synonym 3-Amino-5-(4-fluorophenyl)thiophene-2-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.