Alfa Aesar™ Methyl 3-amino-4-methylthiophene-2-carboxylate, 98%
Manufacturer:  Alfa Aesar™ A1011506
 View more versions of this product
Catalog No. AAA1011506
Specifications
Specifications
Methyl 3-amino-4-methylthiophene-2-carboxylate
84°C to 88°C
85006-31-1
MFCD00051822
methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
CC1=CSC(=C1N)C(=O)OC
171.214
171.22
>100°C (212°F)
5g
C7H9NO2S
973196
YICRPERKKBDRSP-UHFFFAOYSA-N
methyl 3-amino-4-methylthiophene-2-carboxylate
123584
98%
Chemical Identifiers
85006-31-1
171.214
YICRPERKKBDRSP-UHFFFAOYSA-N
123584
CC1=CSC(=C1N)C(=O)OC
C7H9NO2S
MFCD00051822
methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
methyl 3-amino-4-methylthiophene-2-carboxylate
SDS Documents