Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-4-methylthiophene-2-carboxylate, 98%

$95.12 - $1,069.80

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
CAS 85006-31-1
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00051822
Beilstein 973196
Synonym 3-Amino-4-methylthiophene-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1011506

 
alfa aesar™
A1011506
5g Each for $95.12

AAA1011514

 
alfa aesar™
A1011514
25g Each for $335.57

AAA1011522

 
alfa aesar™
A1011522
100g Each for $1,069.80
Description & Specifications

Specifications

Formula Weight 171.22
Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
Assay 98%
Molecular Formula C7H9NO2S
MDL Number MFCD00051822
CAS 85006-31-1
Beilstein 973196
Synonym 3-Amino-4-methylthiophene-2-carboxylic acid methyl ester
Melting Point 84° to 88°C
Quantity 5g
Flash Point >100°C (212°F)