Please login to your online account to display your discounted pricing

Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-4-methyl-2-thiophenecarboxylate
Assay Percent Range 98%
Molecular Formula C7H9NO2S
Formula Weight 171.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC450770050

  SDS
thermo fisher scientific
AC450770050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450770250

  SDS
thermo fisher scientific
AC450770250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.22
Quantity 5g
Chemical Name or Material Methyl 3-amino-4-methyl-2-thiophenecarboxylate
Assay Percent Range 98%
CAS 85006-31-1
Packaging Glass bottle
Molecular Formula C7H9NO2S