Learn More
Methyl 3-amino-4-hydroxybenzoate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6689406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 3-amino-4-hydroxybenzoate | |
141°C to 143°C | |
MFCD00017095 | |
5424619 | |
VNQABZCSYCTZMS-UHFFFAOYSA-N | |
methyl 3-amino-4-hydroxybenzoate | |
10815 | |
167.16 | |
Solid |
536-25-4 | |
C8H9NO3 | |
5 g | |
orthocaine, orthoform, aminobenz, orthoderm, 3-amino-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-amino-4-hydroxy-, methyl ester, methyl m-amino-p-hydroxybenzoate, methyl 4-hydroxy-3-aminobenzoate, methyl 3-amino-4-hydroxybenzenecarboxylate, unii-jk16yi13qk | |
COC(=O)C1=CC(=C(C=C1)O)N | |
167.164 | |
CHEBI:34904 | |
95% |
Chemical Identifiers
536-25-4 | |
167.164 | |
VNQABZCSYCTZMS-UHFFFAOYSA-N | |
10815 | |
methyl 3-amino-4-hydroxybenzoate |
C8H9NO3 | |
MFCD00017095 | |
orthocaine, orthoform, aminobenz, orthoderm, 3-amino-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-amino-4-hydroxy-, methyl ester, methyl m-amino-p-hydroxybenzoate, methyl 4-hydroxy-3-aminobenzoate, methyl 3-amino-4-hydroxybenzenecarboxylate, unii-jk16yi13qk | |
CHEBI:34904 | |
COC(=O)C1=CC(=C(C=C1)O)N |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-627-3
RTECSNumber : DG2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only