missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Amino-4-chlorobenzoate, TCI America™

$76.39 - $229.17

Chemical Identifiers

CAS 40872-87-5
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
InChI Key LOCJPOYKBUUVKU-UHFFFAOYSA-N
Synonym 3-Amino-4-chlorobenzoic Acid Methyl Ester
PubChem CID 593055
IUPAC Name methyl 3-amino-4-chlorobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M177825G View Documents TCI America
M177825G
25g Each for $229.17
 
M17785G View Documents TCI America
M17785G
5g Each for $76.39
 
Specifications

Chemical Identifiers

40872-87-5
185.607
3-Amino-4-chlorobenzoic Acid Methyl Ester
methyl 3-amino-4-chlorobenzoate
C8H8ClNO2
LOCJPOYKBUUVKU-UHFFFAOYSA-N
593055
COC(=O)C1=CC(=C(C=C1)Cl)N

Specifications

Yellow
25g
C8H8ClNO2
LOCJPOYKBUUVKU-UHFFFAOYSA-N
methyl 3-amino-4-chlorobenzoate
593055
Crystal-Powder at 20°C
Methyl 3-Amino-4-chlorobenzoate
83°C
40872-87-5
3-Amino-4-chlorobenzoic Acid Methyl Ester
COC(=O)C1=CC(=C(C=C1)Cl)N
185.607
185.61
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1481

TSCA : No