missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-amino-2-methylbenzoate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5557906

 View more versions of this product

Catalog No. AAH5557906


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl 3-amino-2-methylbenzoate
Colorless to Brown
300°C
C9H11NO2
MFCD00211328
3-amino-2-methylbenzoic acid methyl ester, methyl3-amino-2-methylbenzoate, methyl-2-methyl-3-aminobenzoate, methyl 3-amino-2-methyl-benzoate, methyl2-methyl-3-aminobenzoate, 3-amino-o-toluic acid methyl ester, benzoic acid, 3-amino-2-methyl-, methyl ester, pubchem4978, acmc-1c8sv, ksc495o4h
CC1=C(C=CC=C1N)C(=O)OC
165.192
165.19
Liquid
18583-89-6
1.146 g/mL
104°C (219°F)
1.573
5 g
ZOOQFAUFPWXUMI-UHFFFAOYSA-N
methyl 3-amino-2-methylbenzoate
3801025
96%

Chemical Identifiers

18583-89-6
165.192
ZOOQFAUFPWXUMI-UHFFFAOYSA-N
3801025
CC1=C(C=CC=C1N)C(=O)OC
C9H11NO2
MFCD00211328
3-amino-2-methylbenzoic acid methyl ester, methyl3-amino-2-methylbenzoate, methyl-2-methyl-3-aminobenzoate, methyl 3-amino-2-methyl-benzoate, methyl2-methyl-3-aminobenzoate, 3-amino-o-toluic acid methyl ester, benzoic acid, 3-amino-2-methyl-, methyl ester, pubchem4978, acmc-1c8sv, ksc495o4h
methyl 3-amino-2-methylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions