missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-amino-2-methylbenzoate, 96%

Manufacturer:  Alfa Aesar™ H5557914

 View more versions of this product

Catalog No. AAH5557914



Specifications

Specifications

Methyl 3-amino-2-methylbenzoate
104°C (219°F)
300°C
25g
C9H11NO2
3-amino-2-methylbenzoic acid methyl ester
CC1=C(C=CC=C1N)C(=O)OC
165.192
165.19
96%
1.146 g/mL
1.573
Colorless to Brown
18583-89-6
MFCD00211328
ZOOQFAUFPWXUMI-UHFFFAOYSA-N
methyl 3-amino-2-methylbenzoate
3801025
Liquid

Chemical Identifiers

18583-89-6
165.192
ZOOQFAUFPWXUMI-UHFFFAOYSA-N
3801025
CC1=C(C=CC=C1N)C(=O)OC
C9H11NO2
MFCD00211328
3-amino-2-methylbenzoic acid methyl ester
methyl 3-amino-2-methylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents