Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-dinitrobenzoate, 98%

$36.16 - $304.36

Chemical Identifiers

Chemical Name or Material Methyl 3,5-dinitrobenzoate
CAS 2702-58-1
Molecular Formula C8H6N2O6
Formula Weight 226.15
MDL Number MFCD00017016
Beilstein 1252593
Synonym 3,5-Dinitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2297309

  SDS
alfa aesar™
B2297309
10g Each for $36.16

AAB2297322

  SDS
alfa aesar™
B2297322
100g Each for $136.97

AAB2297330

  SDS
alfa aesar™
B2297330
250g Each for $304.36
Description & Specifications

Specifications

Formula Weight 226.15
Quantity 10g
MDL Number MFCD00017016
Beilstein 1252593
CAS 2702-58-1
Molecular Formula C8H6N2O6
Assay 98%
Chemical Name or Material Methyl 3,5-dinitrobenzoate
Synonym 3,5-Dinitrobenzoic acid methyl ester
Melting Point 108° to 112°C