missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3,5-Dihydroxy-4-methoxybenzoate, TCI America™

$404.88 - $1,423.92

Chemical Identifiers

CAS 24093-81-0
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00017195
InChI Key SUGIJIFASYORQN-UHFFFAOYSA-N
Synonym 3,5-Dihydroxy-4-methoxybenzoic Acid Methyl Ester
PubChem CID 5319726
IUPAC Name methyl 3,5-dihydroxy-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1O)C(=O)OC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27941G View Documents TCI America
M27941G
1g Each for $404.88
 
M27945G View Documents TCI America
M27945G
5g Each for $1,423.92
 
Specifications

Chemical Identifiers

24093-81-0
198.174
SUGIJIFASYORQN-UHFFFAOYSA-N
5319726
COC1=C(C=C(C=C1O)C(=O)OC)O
C9H10O5
MFCD00017195
3,5-Dihydroxy-4-methoxybenzoic Acid Methyl Ester
methyl 3,5-dihydroxy-4-methoxybenzoate

Specifications

White
1g
C9H10O5
3,5-Dihydroxy-4-methoxybenzoic Acid Methyl Ester
COC1=C(C=C(C=C1O)C(=O)OC)O
198.174
198.17
≥98.0% (GC)
149°C
24093-81-0
MFCD00017195
SUGIJIFASYORQN-UHFFFAOYSA-N
methyl 3,5-dihydroxy-4-methoxybenzoate
5319726
Crystal-Powder at 20°C
Methyl 3,5-Dihydroxy-4-methoxybenzoate
Safety and Handling

Safety and Handling

TSCA : No