Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-dichlorobenzoate, 98%

$304.78 - $304.78

Chemical Identifiers

Chemical Name or Material Methyl 3,5-dichlorobenzoate
CAS 2905-67-1
Molecular Formula C8H6Cl2O2
Formula Weight 205.04
MDL Number MFCD00032710
Synonym 3,5-Dichlorobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2531114

 
alfa aesar™
B2531114
25g Each for $304.78
Description & Specifications

Specifications

Formula Weight 205.04
Chemical Name or Material Methyl 3,5-dichlorobenzoate
Assay 98%
Molecular Formula C8H6Cl2O2
MDL Number MFCD00032710
CAS 2905-67-1
Synonym 3,5-Dichlorobenzoic acid methyl ester
Health Hazard 1 Warning
Melting Point 58° to 60°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.