Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-dibromo-4-methylbenzoate, 98%

$60.66 - $250.91

Chemical Identifiers

Chemical Name or Material Methyl 3,5-dibromo-4-methylbenzoate
CAS 74896-66-5
Molecular Formula C9H8Br2O2
Formula Weight 307.98
MDL Number MFCD00010729
Beilstein 2580945
Synonym 3,5-Dibromo-4-methylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2534806

  SDS
alfa aesar™
B2534806
5g Each for $60.66

AAB2534814

  SDS
alfa aesar™
B2534814
25g Each for $250.91
Description & Specifications

Specifications

Formula Weight 307.98
Chemical Name or Material Methyl 3,5-dibromo-4-methylbenzoate
Assay 98%
Molecular Formula C9H8Br2O2
MDL Number MFCD00010729
CAS 74896-66-5
Beilstein 2580945
Synonym 3,5-Dibromo-4-methylbenzoic acid methyl ester
Melting Point 86° to 90°C
Quantity 5g