Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate, 99%

$57.58 - $197.03

Chemical Identifiers

Chemical Name or Material Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
CAS 715-33-3
Molecular Formula C9H8Br2O4
Formula Weight 339.98
MDL Number MFCD00082715
Beilstein 2116569
Synonym 3,5-Dibromo-2,4-dihydroxy-6-methylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1209803

 
alfa aesar™
L1209803
1g Each for $57.58

AAL1209806

 
alfa aesar™
L1209806
5g Each for $197.03
Description & Specifications

Specifications

Formula Weight 339.98
Chemical Name or Material Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
MDL Number MFCD00082715
Assay 99%
Molecular Formula C9H8Br2O4
CAS 715-33-3
Synonym 3,5-Dibromo-2,4-dihydroxy-6-methylbenzoic acid methyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2116569
Melting Point 104° to 107°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.