missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate, 99%

Manufacturer:  Alfa Aesar™ L1209803

 View more versions of this product

Catalog No. AAL1209803



Specifications

Specifications

Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
1g
C9H8Br2O4
2116569
OKALYLLBFOYLQB-UHFFFAOYSA-N
methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
4121774
99%
104°C to 107°C
715-33-3
MFCD00082715
3,5-dibromo-2,4-dihydroxy-6-methylbenzoic acid methyl ester, methyl3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
CC1=C(C(=C(C(=C1Br)O)Br)O)C(=O)OC
339.967
339.98

Chemical Identifiers

715-33-3
339.967
OKALYLLBFOYLQB-UHFFFAOYSA-N
4121774
CC1=C(C(=C(C(=C1Br)O)Br)O)C(=O)OC
C9H8Br2O4
MFCD00082715
3,5-dibromo-2,4-dihydroxy-6-methylbenzoic acid methyl ester, methyl3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents