missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3,5-Dibenzyloxybenzoate, TCI America™

$187.92 - $647.80

Chemical Identifiers

CAS 58605-10-0
Molecular Formula C22H20O4
Molecular Weight (g/mol) 348.398
MDL Number MFCD00157205
InChI Key GBQCMRLPXFXVIN-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester
PubChem CID 2733652
IUPAC Name methyl 3,5-bis(phenylmethoxy)benzoate
SMILES COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20925G View Documents TCI America
B20925G
5g Each for $187.92
 
B209225G View Documents TCI America
B209225G
25g Each for $647.80
 
Specifications

Chemical Identifiers

58605-10-0
348.398
GBQCMRLPXFXVIN-UHFFFAOYSA-N
2733652
COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
C22H20O4
MFCD00157205
3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester
methyl 3,5-bis(phenylmethoxy)benzoate

Specifications

White-Yellow
5g
C22H20O4
3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester
COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3
348.398
348.40
≥98.0% (GC)
71°C
58605-10-0
MFCD00157205
GBQCMRLPXFXVIN-UHFFFAOYSA-N
methyl 3,5-bis(phenylmethoxy)benzoate
2733652
Crystal-Powder at 20°C
Methyl 3,5-Dibenzyloxybenzoate
Safety and Handling

Safety and Handling

TSCA : No