missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3,5-Dibenzyloxybenzoate 98.0+%, TCI America™
Supplier: TCI America B209225G
Specifications
Methyl 3,5-Dibenzyloxybenzoate | |
71°C | |
C22H20O4 | |
25 g | |
GBQCMRLPXFXVIN-UHFFFAOYSA-N | |
methyl 3,5-bis(phenylmethoxy)benzoate | |
2733652 | |
≥98.0% (GC) |
58605-10-0 | |
White-Yellow | |
MFCD00157205 | |
3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester | |
COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3 | |
348.398 | |
348.40 | |
Crystalline Powder |
Chemical Identifiers
58605-10-0 | |
348.398 | |
GBQCMRLPXFXVIN-UHFFFAOYSA-N | |
2733652 | |
COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3 |
C22H20O4 | |
MFCD00157205 | |
3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester | |
methyl 3,5-bis(phenylmethoxy)benzoate |
Safety and Handling
TSCA : No