missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate, 98%

Manufacturer:  Alfa Aesar™ H5565703

 View more versions of this product

Catalog No. AAH5565703



Specifications

Specifications

Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
1g
C6H7ClN4O2
3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester
COC(=O)C1=C(N=C(C(=N1)Cl)N)N
202.598
202.6
211°C to 215°C
1458-01-1
MFCD01928388
KOOBYHRLTYIPTH-UHFFFAOYSA-N
methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
73827
98%

Chemical Identifiers

1458-01-1
202.598
KOOBYHRLTYIPTH-UHFFFAOYSA-N
73827
COC(=O)C1=C(N=C(C(=N1)Cl)N)N
C6H7ClN4O2
MFCD01928388
3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester
methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P314-P330-P501c

H301-H319-H335-H373

EINECSNumber : 215-946-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents