Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,5-bis(trifluoromethyl)benzoate, 98%

$44.78 - $166.24

Chemical Identifiers

Chemical Name or Material Methyl 3,5-bis(trifluoromethyl)benzoate
CAS 26107-80-2
Molecular Formula C10H6F6O2
Formula Weight 272.15
MDL Number MFCD00052317
Synonym 3,5-Bis(trifluoromethyl)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2099003

 
alfa aesar™
B2099003
1g Each for $44.78

AAB2099006

 
alfa aesar™
B2099006
5g Each for $166.24
Description & Specifications

Specifications

Formula Weight 272.15
Chemical Name or Material Methyl 3,5-bis(trifluoromethyl)benzoate
Assay 98%
Molecular Formula C10H6F6O2
MDL Number MFCD00052317
CAS 26107-80-2
Synonym 3,5-Bis(trifluoromethyl)benzoic acid methyl ester
Health Hazard 1 Warning
Boiling Point 85° to 87°C (15mmHg)
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.