missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%

Manufacturer:  Alfa Aesar™ H5577014

 View more versions of this product

Catalog No. AAH5577014



Specifications

Specifications

Methyl 3-(4-methyl-1-piperazinyl)benzoate
1.5630
Colorless to Yellow
474334-89-9
MFCD07369792
LPOJJZXQESTVEV-UHFFFAOYSA-N
methyl 3-(4-methylpiperazin-1-yl)benzoate
16217966
Liquid
1.119g/mL
169°C to 172°C (0.9mmHg)
25g
C13H18N2O2
1-3-carbomethoxyphenyl-4-methylpipera&
CN1CCN(CC1)C2=CC=CC(=C2)C(=O)OC
234.299
234.3
97%

Chemical Identifiers

474334-89-9
234.299
LPOJJZXQESTVEV-UHFFFAOYSA-N
16217966
CN1CCN(CC1)C2=CC=CC(=C2)C(=O)OC
C13H18N2O2
MFCD07369792
1-3-carbomethoxyphenyl-4-methylpipera&
methyl 3-(4-methylpiperazin-1-yl)benzoate
Documents