missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-(4-Hydroxy-3-methoxyphenyl)propionate, TCI America™
$311.67 - $1,074.05
Chemical Identifiers
CAS | 56024-44-3 |
---|---|
Molecular Formula | C11H14O4 |
Molecular Weight (g/mol) | 210.229 |
InChI Key | PDTCYIZPTRRYOT-UHFFFAOYSA-N |
Synonym | 3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester |
PubChem CID | 523498 |
IUPAC Name | methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate |
SMILES | COC1=C(C=CC(=C1)CCC(=O)OC)O |
Chemical Identifiers
56024-44-3 | |
210.229 | |
3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester | |
methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate |
C11H14O4 | |
PDTCYIZPTRRYOT-UHFFFAOYSA-N | |
523498 | |
COC1=C(C=CC(=C1)CCC(=O)OC)O |
Specifications
146°C | |
5g | |
C11H14O4 | |
PDTCYIZPTRRYOT-UHFFFAOYSA-N | |
methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate | |
523498 | |
Clear Liquid at 20°C | |
Methyl 3-(4-Hydroxy-3-methoxyphenyl)propionate |
Yellow | |
56024-44-3 | |
3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester | |
COC1=C(C=CC(=C1)CCC(=O)OC)O | |
210.229 | |
210.23 | |
≥98.0% (GC) |
Safety and Handling
TSCA : No