Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,4-dimethylbenzoate, 98%

$1,898.51 - $1,898.51

Chemical Identifiers

Chemical Name or Material Methyl 3,4-dimethylbenzoate
CAS 38404-42-1
Molecular Formula C10H12O2
Formula Weight 164.21
MDL Number MFCD00219861
Synonym 3,4-Dimethylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2002430

  SDS
alfa aesar™
B2002430
250g Each for $1,898.51
Description & Specifications

Specifications

Formula Weight 164.21
Assay 98%
CAS 38404-42-1
Molecular Formula C10H12O2
MDL Number MFCD00219861
Melting Point 35° to 37°C
Quantity 250g
Synonym 3,4-Dimethylbenzoic acid methyl ester
Chemical Name or Material Methyl 3,4-dimethylbenzoate

Applications
It is used to produce 3,4-Dimethyl-benzyl alcohol.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.