Please login to your online account to display your discounted pricing

Methyl 3,4-dimethoxybenzoate, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3, 4-dimethoxybenzoate
Name Note 98+%
CAS 2150-38-1
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C10H12O4
Linear Formula (CH3O)2C6H3CO2CH3
Formula Weight 196.2
MDL Number MFCD00008430
Synonym Methyl veratrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189230500

  SDS
thermo fisher scientific
189230500
Plastic bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Granular Powder
Formula Weight 196.2
Quantity 50g
Color Brown
EINECS Number 218-424-1
TSCA TSCA
Melting Point 58° to 62°C
Beilstein 10, 396
Chemical Name or Material Methyl 3, 4-dimethoxybenzoate
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C10H12O4
Linear Formula (CH3O)2C6H3CO2CH3
MDL Number MFCD00008430
CAS 2150-38-1
Synonym Methyl veratrate
Infrared Spectrum Authentic
Packaging Plastic bottle