Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,4-dimethoxybenzoate, 98+%

$62.34 - $252.45

Chemical Identifiers

Chemical Name or Material Methyl 3,4-dimethoxybenzoate
CAS 2150-38-1
Molecular Formula C10H12O4
Formula Weight 196.2
MDL Number MFCD00008430
Beilstein 2215244
Synonym 3,4-Dimethoxybenzoic acid methyl ester; Methyl veratrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2356818

 
alfa aesar™
B2356818
50g Each for $62.34

AAB2356830

 
alfa aesar™
B2356830
250g Each for $252.45
Description & Specifications

Specifications

Formula Weight 196.2
Chemical Name or Material Methyl 3,4-dimethoxybenzoate
TSCA TSCA
Assay ≥98%
Molecular Formula C10H12O4
MDL Number MFCD00008430
CAS 2150-38-1
Beilstein 2215244
Synonym 3,4-Dimethoxybenzoic acid methyl ester; Methyl veratrate
Melting Point 57° to 61°C
Boiling Point 283°C
Quantity 50g