Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,4-difluorobenzoate, 97%

$108.64 - $108.64

Chemical Identifiers

Chemical Name or Material Methyl 3,4-difluorobenzoate
CAS 369-25-5
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD00276597
Synonym 3,4-Difluorobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1986606

  SDS
alfa aesar™
A1986606
5g Each for $108.64
Description & Specifications

Specifications

Formula Weight 172.13
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3,4-Difluorobenzoic acid methyl ester
Boiling Point 87° to 89°C (15mmHg)
Molecular Formula C8H6F2O2
Assay 97%
Chemical Name or Material Methyl 3,4-difluorobenzoate
MDL Number MFCD00276597
Health Hazard 1 Warning
CAS 369-25-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.