missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3,4-Dichlorophenylacetate, TCI America™

$106.95 - $366.67

Chemical Identifiers

CAS 6725-44-6
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00800680
InChI Key FWVOAIHAIWHHDM-UHFFFAOYSA-N
Synonym methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
PubChem CID 1268257
IUPAC Name methyl 2-(3,4-dichlorophenyl)acetate
SMILES COC(=O)CC1=CC(=C(C=C1)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25575G View Documents TCI America
M25575G
5g Each for $106.95
 
M255725G View Documents TCI America
M255725G
25g Each for $366.67
 
Specifications

Chemical Identifiers

6725-44-6
219.061
FWVOAIHAIWHHDM-UHFFFAOYSA-N
1268257
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
C9H8Cl2O2
MFCD00800680
methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
methyl 2-(3,4-dichlorophenyl)acetate

Specifications

Yellow
6725-44-6
MFCD00800680
FWVOAIHAIWHHDM-UHFFFAOYSA-N
methyl 2-(3,4-dichlorophenyl)acetate
1268257
Clear Liquid at 20°C
Methyl 3,4-Dichlorophenylacetate
5g
C9H8Cl2O2
methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester
COC(=O)CC1=CC(=C(C=C1)Cl)Cl
219.061
219.06
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No