Please login to your online account to display your discounted pricing

Methyl 3,4-dichlorophenylacetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3, 4-dichlorophenylacetate
Name Note 99%
CAS 6725-44-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8Cl2O2
Formula Weight 219.07
MDL Number MFCD00800680
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351080250

  SDS
thermo fisher scientific
351080250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 219.07
Physical Form Liquid After Melting
Quantity 25g
Color Clear colorless to light yellow
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Name Note 99%
Molecular Formula C9H8Cl2O2
Chemical Name or Material Methyl 3, 4-dichlorophenylacetate
MDL Number MFCD00800680
Assay Percent Range 99%
CAS 6725-44-6