missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3,4-dichlorophenylacetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 351080250
Specifications
Methyl 3, 4-dichlorophenylacetate | |
6725-44-6 | |
98.5% min. (GC) | |
C9H8Cl2O2 | |
MFCD00800680 | |
methyl 2-3,4-dichlorophenyl acetate, methyl 3,4-dichlorophenylacetate, 3,4-dichlorophenylacetic acid methyl ester, 3,4-dichloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 3,4-dichloro-, methyl ester, pubchem20251, acmc-1bhwn, methyl 3,4-dichloro-phenylacetate, methyl 2-3,4-dichlorophenyl ethanoate, 3,4 dichlorophenyl acetic acid methyl ester | |
COC(=O)CC1=CC(=C(C=C1)Cl)Cl | |
219.07 | |
219.07 |
99% | |
Authentic | |
Glass bottle | |
1.5400 to 1.5420 | |
25g | |
FWVOAIHAIWHHDM-UHFFFAOYSA-N | |
methyl 2-(3,4-dichlorophenyl)acetate | |
1268257 | |
99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only