Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3,4-dichlorophenylacetate, 97+%

$34.35 - $483.68

Chemical Identifiers

Chemical Name or Material Methyl 3,4-dichlorophenylacetate
CAS 6725-44-6
Molecular Formula C9H8Cl2O2
Formula Weight 219.07
MDL Number MFCD00800680
Synonym 3,4-Dichlorophenylacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2151804

  SDS
alfa aesar™
B2151804
2g Each for $34.35

AAB2151809

  SDS
alfa aesar™
B2151809
10g Each for $119.93

AAB2151818

  SDS
alfa aesar™
B2151818
50g Each for $483.68
Description & Specifications

Specifications

Formula Weight 219.07
CAS 6725-44-6
Molecular Formula C9H8Cl2O2
Chemical Name or Material Methyl 3,4-dichlorophenylacetate
Quantity 2g
MDL Number MFCD00800680
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3,4-Dichlorophenylacetic acid methyl ester
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
Health Hazard 1 Warning
Melting Point 27° to 30°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.