missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,4-Diaminobenzoate 98.0+%, TCI America™
Supplier: TCI America M234625G
Specifications
| Methyl 3,4-Diaminobenzoate | |
| 109°C | |
| MFCD00017098 | |
| 3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate | |
| COC(=O)C1=CC=C(N)C(N)=C1 | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
| 36692-49-6 | |
| C8H10N2O2 | |
| 25 g | |
| IOPLHGOSNCJOOO-UHFFFAOYSA-N | |
| methyl 3,4-diaminobenzoate | |
| 135524 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 36692-49-6 | |
| 166.18 | |
| IOPLHGOSNCJOOO-UHFFFAOYSA-N | |
| 135524 | |
| COC(=O)C1=CC=C(N)C(N)=C1 |
| C8H10N2O2 | |
| MFCD00017098 | |
| 3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate | |
| methyl 3,4-diaminobenzoate |
Safety and Handling
TSCA : No