Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride, 97%

$98.84 - $355.59

Chemical Identifiers

Chemical Name or Material Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride
CAS 103565-40-8
Molecular Formula C11H15NO2·HCl
Formula Weight 229.71
MDL Number MFCD11113035
Beilstein 14326402
Synonym 3-[4-(Aminomethyl)phenyl]propionic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5103103

  SDS
alfa aesar™
H5103103
1g Each for $355.59

AAH51031MD

  SDS
alfa aesar™
H51031MD
250mg Each for $98.84
Description & Specifications

Specifications

Formula Weight 229.71
Chemical Name or Material Methyl 3-[4-(aminomethyl)phenyl]propionate hydrochloride
Health Hazard 1 Warning
Synonym 3-[4-(Aminomethyl)phenyl]propionic acid methyl ester hydrochloride
Quantity 1g
Assay 97%
MDL Number MFCD11113035
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H15NO2·HCl
Beilstein 14326402
CAS 103565-40-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.