missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3,4,5-trimethoxybenzoate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1193322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 3,4,5-trimethoxybenzoate | |
81°C to 85°C | |
C11H14O5 | |
100 g | |
trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8 | |
COC1=CC(=CC(=C1OC)OC)C(=O)OC | |
226.228 | |
226.23 |
1916-07-0 | |
274°C to 275°C | |
MFCD00008431 | |
2218156 | |
KACHFMOHOPLTNX-UHFFFAOYSA-N | |
methyl 3,4,5-trimethoxybenzoate | |
15956 | |
≥98% |
Chemical Identifiers
1916-07-0 | |
226.228 | |
KACHFMOHOPLTNX-UHFFFAOYSA-N | |
15956 | |
COC1=CC(=CC(=C1OC)OC)C(=O)OC |
C11H14O5 | |
MFCD00008431 | |
trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8 | |
methyl 3,4,5-trimethoxybenzoate |
Safety and Handling
EINECSNumber : 217-629-3
RTECSNumber : DI0820000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only