missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98.0+%, TCI America™
$64.70 - $189.13
Chemical Identifiers
CAS | 480425-35-2 |
---|---|
Molecular Formula | C14H19BO4 |
Molecular Weight (g/mol) | 262.112 |
MDL Number | MFCD03789258 |
InChI Key | JBJGSVBGUBATNH-UHFFFAOYSA-N |
Synonym | 3-methoxycarbonylphenylboronic acid pinacol ester, methyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-methoxycarbonylphenylboronic acid, pinacol ester, 3-methoxycarbonyl phenylboronic acid pinacol ester, methyl 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, pubchem9527, 3-carbomethoxyphenylboronic acid pinacol ester, 3-methoxycarbonyl phenyl boronic acid pinacol ester |
PubChem CID | 2734651 |
IUPAC Name | methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)OC |
Chemical Identifiers
480425-35-2 | |
262.112 | |
JBJGSVBGUBATNH-UHFFFAOYSA-N | |
2734651 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)OC |
C14H19BO4 | |
MFCD03789258 | |
3-methoxycarbonylphenylboronic acid pinacol ester, methyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-methoxycarbonylphenylboronic acid, pinacol ester, 3-methoxycarbonyl phenylboronic acid pinacol ester, methyl 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, pubchem9527, 3-carbomethoxyphenylboronic acid pinacol ester, 3-methoxycarbonyl phenyl boronic acid pinacol ester | |
methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
Specifications
480425-35-2 | |
White | |
MFCD03789258 | |
3-methoxycarbonylphenylboronic acid pinacol ester, methyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-methoxycarbonylphenylboronic acid, pinacol ester, 3-methoxycarbonyl phenylboronic acid pinacol ester, methyl 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, pubchem9527, 3-carbomethoxyphenylboronic acid pinacol ester, 3-methoxycarbonyl phenyl boronic acid pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)OC | |
262.112 | |
262.11 | |
Crystalline Powder |
95°C | |
C14H19BO4 | |
1 g | |
JBJGSVBGUBATNH-UHFFFAOYSA-N | |
methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate | |
2734651 | |
≥98.0% (GC,T) | |
Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
Safety and Handling
TSCA : No