Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-(3-pyridyl)propionate, 98%

$54.39 - $167.26

Chemical Identifiers

Chemical Name or Material Methyl 3-(3-pyridyl)propionate
CAS 84199-98-4
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD01075660
Beilstein 132852
Synonym 3-(3-Pyridyl)propionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1459006

  SDS
alfa aesar™
L1459006
5g Each for $167.26

AAL1459003

  SDS
alfa aesar™
L1459003
1g Each for $54.39
Description & Specifications

Specifications

Formula Weight 165.19
MDL Number MFCD01075660
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 134°C (12mmHg)
Chemical Name or Material Methyl 3-(3-pyridyl)propionate
CAS 84199-98-4
Health Hazard 1 Warning
Assay 98%
Synonym 3-(3-Pyridyl)propionic acid methyl ester
Quantity 5g
Beilstein 132852
Molecular Formula C9H11NO2
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.